Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01764627_m1
          See other LINGO2 GE Assays ›
          Gene Symbol:
          Lingo2
          Gene Name:
          leucine rich repeat and Ig domain containing 2
          Gene Aliases:
          RGD1310773
          Chromosome Location:
          Chr.5: 51466367 - 51638161 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01764627_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lingo2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107926.1 NP_001101396.1 304 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002514.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001107926.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P081817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73662 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49732 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130125 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112990 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081817 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P081817 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P532307 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P403260 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87619 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42428 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875975 - -
          Affymetrix Rat Gene 1.1 ST Array 10875974 - -
          Rat Genome 230 2.0 Array 1393841_at - -
          Rat Genome 230 2.0 Array 1376787_PM_at - -
          Rat Genome 230 2.0 Array 1393841_PM_at - -
          Rat Genome 230 2.0 Array 1376787_at - -
          Affymetrix HT Rat Focus Array 1393841_PM_at - -
          Affymetrix HT Rat Focus Array 1376787_PM_at - -
          Affymetrix HT Rat Focus Array 1393841_at - -
          Affymetrix HT Rat Focus Array 1376787_at - -
          Affymetrix Rat Clariom S Assay PSR0500018577.rn.1 TC0500002514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500018590.rn.1 TC0500002514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500018578.rn.1 TC0500002514.rn.1 RTA TC
          Rat Expression Set 230 1393841_at - -
          Rat Expression Set 230 1376787_PM_at - -
          Rat Expression Set 230 1376787_at - -
          Rat Expression Set 230 1393841_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10875974 - -
          Rat Genome U34 Set rc_AA859317_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21520960 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline