Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01765043_g1
          See other PRKCQ GE Assays ›
          Gene Symbol:
          Prkcq
          Gene Name:
          protein kinase C, theta
          Gene Aliases:
          Pkcq
          Chromosome Location:
          Chr.17: 70971915 - 71105286 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01765043_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkcq
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001276721.1 NP_001263650.1 17 - 18 2147 69
          GenBank mRNA AB020614.1 - 3 - 4 394 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700002081.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Apoptosis Protein Kinase 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          immune system process
          regulation of transcription, DNA-templated
          protein phosphorylation
          membrane protein ectodomain proteolysis
          inflammatory response
          aging
          response to heat
          response to glucose
          regulation of G2/M transition of mitotic cell cycle
          response to organic cyclic compound
          regulation of vasoconstriction
          neuron differentiation
          positive regulation of interleukin-17 production
          positive regulation of interleukin-4 production
          response to insulin
          intracellular signal transduction
          positive regulation of T cell proliferation
          tissue regeneration
          positive regulation of NF-kappaB import into nucleus
          positive regulation of interleukin-2 biosynthetic process
          respiratory burst
          negative regulation of insulin receptor signaling pathway
          positive regulation of protein secretion
          positive regulation of T cell activation
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of filopodium assembly
          positive regulation of stress fiber assembly
          negative regulation of T cell apoptotic process
          regulation of platelet aggregation
          positive regulation of T-helper 17 type immune response
          positive regulation of T-helper 2 cell activation
          protein serine/threonine kinase activity
          protein kinase C activity
          protein binding
          ATP binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P608713 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16037 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700523938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92737 - -
          Agilent Rat cDNA Microarray (G4105A) 700949956 - -
          Agilent Rat cDNA Microarray (G4105A) 700523938 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10799439 - -
          Affymetrix Rat Gene 1.0 ST Array 10799437 - -
          Affymetrix Rat Gene 1.0 ST Array 10799438 - -
          Affymetrix Rat Gene 1.1 ST Array 10799435 - -
          Rat Genome 230 2.0 Array 1376259_PM_at - -
          Rat Genome 230 2.0 Array 1376259_at - -
          Affymetrix HT Rat Focus Array 1376259_at - -
          Affymetrix HT Rat Focus Array 1376259_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700014306.rn.1 TC1700002081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014305.rn.1 TC1700002081.rn.1 RTA TC
          Rat Expression Set 230 1376259_at - -
          Rat Expression Set 230 1376259_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10799435 - -
          Rat Genome U34 Set rc_AI171093_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22206538 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368580 - -
          RatRef-12 Expression BeadChip ILMN_69891 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline