Search
Search
| Gene Symbol: Katna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004217.4 | NP_001004217.2 | 6 - 7 | 927 | 104 | |
| GenBank mRNA | AY621629.1 | - | 5 - 6 | 731 | 104 | |
| BC097929.1 | - | 6 - 7 | 927 | 7444887 | 104 | |
Set Membership: |
|