Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01767120_m1
See other KCNE1 GE Assays ›
Gene Symbol:
Kcne1
Gene Name:
potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 1
Gene Aliases:
Isk, m
Chromosome Location:
Chr.11: 32498260 - 32508420 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01767120_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Kcne1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_006248035.2
XP_006248097.1
1 - 2
193
60
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006248035.2 transcripts.
Set Membership:
Amplicon length less than or equal to 70
Ion Channel
Rattus norvegicus
Inventoried
Probe spans exons
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
epithelial cell maturation
protein N-linked glycosylation
protein O-linked glycosylation
male gonad development
heart contraction
regulation of membrane repolarization
regulation of ventricular cardiac muscle cell membrane repolarization
cellular response to cAMP
potassium ion export
cellular response to acidic pH
potassium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
ventricular cardiac muscle cell action potential
membrane repolarization
membrane repolarization during action potential
membrane repolarization during cardiac muscle cell action potential
regulation of heart rate by cardiac conduction
negative regulation of protein targeting to membrane
regulation of potassium ion transmembrane transport
positive regulation of potassium ion transmembrane transport
positive regulation of voltage-gated calcium channel activity
regulation of delayed rectifier potassium channel activity
negative regulation of voltage-gated potassium channel activity
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
protein binding
potassium channel regulator activity
telethonin binding
protein heterodimerization activity
voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline