Search
Search
| Gene Symbol: Stt3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134749.1 | NP_001128221.1 | 8 - 9 | 877 | 66 | |
| XM_006242808.1 | XP_006242870.1 | 8 - 9 | 922 | 66 | ||
| GenBank mRNA | BC168223.1 | - | 8 - 9 | 848 | 7324965 | 66 |
Set Membership: |
|