Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767602_g1
          See other ALG12 GE Assays ›
          Gene Symbol:
          Alg12
          Gene Name:
          asparagine-linked glycosylation 12, alpha-1,6-mannosyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 129798663 - 129812388 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767602_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alg12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108104.2 NP_001101574.2 9 - 10 1268 63
          GenBank mRNA BC161934.1 - 9 - 10 1266 9022144 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003679.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          glycosyltransferase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein N-linked glycosylation
          dolichol-linked oligosaccharide biosynthetic process
          biological_process
          mannosylation
          mannosyltransferase activity
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555731 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21419 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21419 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P21419 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21419 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103843 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10906064 - -
          Affymetrix Rat Gene 1.0 ST Array 10906061 - -
          Affymetrix Rat Gene 1.0 ST Array 10906058 - -
          Affymetrix Rat Gene 1.0 ST Array 10906059 - -
          Affymetrix Rat Gene 1.0 ST Array 10906062 - -
          Affymetrix Rat Gene 1.0 ST Array 10906057 - -
          Affymetrix Rat Gene 1.0 ST Array 10906056 - -
          Affymetrix Rat Gene 1.0 ST Array 10906055 - -
          Affymetrix Rat Gene 1.0 ST Array 10906063 - -
          Affymetrix Rat Gene 1.0 ST Array 10906060 - -
          Affymetrix Rat Gene 1.1 ST Array 10906054 - -
          Rat Genome 230 2.0 Array 1372902_at - -
          Rat Genome 230 2.0 Array 1372902_PM_at - -
          Affymetrix HT Rat Focus Array 1372902_at - -
          Affymetrix HT Rat Focus Array 1372902_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700029020.rn.1 TC0700003679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700029021.rn.1 TC0700003679.rn.1 RTA TC
          Rat Expression Set 230 1372902_at - -
          Rat Expression Set 230 1372902_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10906054 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21647332 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371307 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline