Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767651_m1
          See other MPPED1 GE Assays ›
          Gene Symbol:
          Mpped1
          Gene Name:
          metallophosphoesterase domain containing 1
          Gene Aliases:
          RGD1308244
          Chromosome Location:
          Chr.7: 124682868 - 124751398 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767651_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mpped1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130569.1 NP_001124041.1 5 - 6 951 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001557.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P930152 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54161 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P930152 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P930152 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P930152 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142675 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P403415 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898197 - -
          Affymetrix Rat Gene 1.0 ST Array 10898201 - -
          Affymetrix Rat Gene 1.0 ST Array 10898199 - -
          Affymetrix Rat Gene 1.0 ST Array 10898198 - -
          Affymetrix Rat Gene 1.0 ST Array 10898202 - -
          Affymetrix Rat Gene 1.0 ST Array 10898200 - -
          Affymetrix Rat Gene 1.1 ST Array 10898196 - -
          Rat Genome 230 2.0 Array 1388968_PM_at - -
          Rat Genome 230 2.0 Array 1396924_at - -
          Rat Genome 230 2.0 Array 1396924_PM_at - -
          Rat Genome 230 2.0 Array 1388968_at - -
          Affymetrix HT Rat Focus Array 1388968_at - -
          Affymetrix HT Rat Focus Array 1388968_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700011836.rn.1 TC0700001557.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700011837.rn.1 TC0700001557.rn.1 RTA TC
          Rat Expression Set 230 1388968_PM_at - -
          Rat Expression Set 230 1388968_at - -
          Rat Expression Set 230 1396924_at - -
          Rat Expression Set 230 1396924_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898196 - -
          Rat Genome U34 Set rc_AI144789_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21186945 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362968 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline