Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767786_g1
          See other RPL8 GE Assays ›
          Gene Symbol:
          Rpl8 LOC100360117 LOC100910370
          Gene Name:
          ribosomal protein L8
          ribosomal protein L8-like
          60S ribosomal protein L8-like
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.7: 118507014 - 118509292 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 3389589 - 3390465 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 117967516 - 117969888 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn01767786_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034916.2 NP_001030088.2 657 100
          GenBank mRNA BC126063.1 - 647 7307697 100
          CF111102.1 - 657 100
          FQ217739.1 - 656 100
          FQ225104.1 - 657 100
          FQ226443.1 - 655 100
          FQ226648.1 - 655 100
          FQ230224.1 - 659 100
          FQ230609.1 - 664 100
          FQ234329.1 - 664 100
          X62145.1 - 657 100
          Gene Symbol: LOC100360117
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002729143.4 XP_002729189.1 1 670 100
          Gene Symbol: LOC100910370
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750368.3 XP_003750416.1 5 - 6 736 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl8
          Click here to find siRNAs for Gene LOC100360117
          Click here to find siRNAs for Gene LOC100910370

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          rRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084455 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752683 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084455 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112774 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112774 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084455 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112774 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084455 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P774045 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700148681 - -
          Agilent Rat cDNA Microarray (G4105A) 700148681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834407 - -
          Affymetrix Rat Gene 1.0 ST Array 10716061 - -
          Affymetrix Rat Gene 1.0 ST Array 10834408 - -
          Affymetrix Rat Gene 1.1 ST Array 10716060 - -
          Affymetrix Rat Gene 1.1 ST Array 10834406 - -
          Rat Genome 230 2.0 Array 1371305_PM_at - -
          Rat Genome 230 2.0 Array 1371305_at - -
          Rat Genome 230 2.0 Array 1384714_x_at - -
          Rat Genome 230 2.0 Array 1384714_PM_x_at - -
          Affymetrix HT Rat Focus Array 1371305_at - -
          Affymetrix HT Rat Focus Array 1371305_PM_at - -
          Rat Expression Set 230 1371305_PM_at - -
          Rat Expression Set 230 1371305_at - -
          Rat Expression Set 230 1384714_x_at - -
          Rat Expression Set 230 1384714_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10834406 - -
          Affymetrix Rat Gene 1.0 ST Array 10716060 - -
          Rat Genome U34 Set rc_AA892582_s_at - -
          Rat Genome U34 Set X62145cds_at - -
          Rat Genome U34 Set X62145cds_g_at - -
          Rat Genome U34 Set rc_AI169389_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22183070 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369431 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline