Search
Search

Xpnpep3| Gene Symbol: Xpnpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130582.2 | NP_001124054.2 | 7 - 8 | 1103 | 121 | |
| XM_006242121.2 | XP_006242183.1 | 6 - 7 | 920 | 121 | ||
| GenBank mRNA | BC168759.1 | - | 7 - 8 | 1103 | 6888763 | 121 |
Set Membership: |
|