Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767942_m1
          See other PTDSS1 GE Assays ›
          Gene Symbol:
          Ptdss1
          Gene Name:
          phosphatidylserine synthase 1
          Gene Aliases:
          PSS-1
          Chromosome Location:
          Chr.7: 71294140 - 71356153 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767942_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptdss1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012113.1 NP_001012113.1 12 - 13 1421 76
          GenBank mRNA BC087129.1 - 12 - 13 1421 7112273 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000921.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptdss1

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylserine biosynthetic process
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041596 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P476353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77141 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P476353 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P476353 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P476353 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041596 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P429364 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78863 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60122 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42979 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 390 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896016 - -
          Affymetrix Rat Gene 1.0 ST Array 10896006 - -
          Affymetrix Rat Gene 1.0 ST Array 10896011 - -
          Affymetrix Rat Gene 1.0 ST Array 10896012 - -
          Affymetrix Rat Gene 1.0 ST Array 10896010 - -
          Affymetrix Rat Gene 1.0 ST Array 10896007 - -
          Affymetrix Rat Gene 1.0 ST Array 10896015 - -
          Affymetrix Rat Gene 1.0 ST Array 10896009 - -
          Affymetrix Rat Gene 1.0 ST Array 10896014 - -
          Affymetrix Rat Gene 1.0 ST Array 10896013 - -
          Affymetrix Rat Gene 1.0 ST Array 10896018 - -
          Affymetrix Rat Gene 1.0 ST Array 10896019 - -
          Affymetrix Rat Gene 1.0 ST Array 10896017 - -
          Affymetrix Rat Gene 1.0 ST Array 10896008 - -
          Affymetrix Rat Gene 1.1 ST Array 10896004 - -
          Rat Genome 230 2.0 Array 1367497_PM_at - -
          Rat Genome 230 2.0 Array 1367497_at - -
          Affymetrix HT Rat Focus Array 1367497_PM_at - -
          Affymetrix HT Rat Focus Array 1367497_at - -
          Affymetrix Rat Clariom S Assay PSR0700007052.rn.1 TC0700000921.rn.1 RTA TC
          Rat Expression Set 230 1367497_PM_at - -
          Rat Expression Set 230 1367497_at - -
          Rat Expression Set 230 1367497_at - -
          Rat Expression Set 230 1367497_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896004 - -
          Rat Genome U34 Set rc_AI177921_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376151 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline