Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01768056_g1
          See other SHMT2 GE Assays ›
          Gene Symbol:
          Shmt2
          Gene Name:
          serine hydroxymethyltransferase 2 (mitochondrial)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 70824719 - 70829824 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01768056_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Shmt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008322.1 NP_001008323.1 9 - 10 1178 96
          XM_006241453.2 XP_006241515.1 9 - 10 1204 96
          GenBank mRNA BC085331.1 - 9 - 10 1178 7106911 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002923.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Shmt2

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycine metabolic process
          glycine biosynthetic process
          L-serine metabolic process
          L-serine biosynthetic process
          one-carbon metabolic process
          positive regulation of cell proliferation
          glycine biosynthetic process from serine
          methylation
          tetrahydrofolate interconversion
          protein homotetramerization
          chromatin binding
          glycine hydroxymethyltransferase activity
          methyltransferase activity
          L-allo-threonine aldolase activity
          amino acid binding
          pyridoxal phosphate binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P501843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P501843 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P501843 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P501843 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501843 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903070 - -
          Affymetrix Rat Gene 1.0 ST Array 10903062 - -
          Affymetrix Rat Gene 1.0 ST Array 10903065 - -
          Affymetrix Rat Gene 1.0 ST Array 10903066 - -
          Affymetrix Rat Gene 1.0 ST Array 10903063 - -
          Affymetrix Rat Gene 1.0 ST Array 10903071 - -
          Affymetrix Rat Gene 1.0 ST Array 10903068 - -
          Affymetrix Rat Gene 1.0 ST Array 10903072 - -
          Affymetrix Rat Gene 1.0 ST Array 10903067 - -
          Affymetrix Rat Gene 1.0 ST Array 10903069 - -
          Affymetrix Rat Gene 1.0 ST Array 10903073 - -
          Affymetrix Rat Gene 1.0 ST Array 10903064 - -
          Affymetrix Rat Gene 1.1 ST Array 10903061 - -
          Rat Genome 230 2.0 Array 1388695_PM_at - -
          Rat Genome 230 2.0 Array 1388695_at - -
          Affymetrix HT Rat Focus Array 1388695_PM_at - -
          Affymetrix HT Rat Focus Array 1388695_at - -
          Affymetrix Rat Clariom S Assay PSR0700022605.rn.1 TC0700002923.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022608.rn.1 TC0700002923.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022607.rn.1 TC0700002923.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022606.rn.1 TC0700002923.rn.1 RTA TC
          Rat Expression Set 230 1388695_PM_at - -
          Rat Expression Set 230 1388695_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903061 - -
          Rat Genome U34 Set rc_AI104298_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21271735 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368671 - -
          RatRef-12 Expression BeadChip ILMN_60571 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline