Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01768657_m1
          See other FGD2 GE Assays ›
          Gene Symbol:
          Fgd2
          Gene Name:
          FYVE, RhoGEF and PH domain containing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 6973355 - 6990023 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01768657_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107617.2 NP_001101087.2 13 - 14 1631 82
          XM_006256205.2 XP_006256267.1 13 - 14 1677 82
          XM_008772729.1 XP_008770951.1 12 - 13 1584 82
          GenBank mRNA BC089841.1 - 13 - 14 1659 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000289.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Process(es)

          Rho guanyl-nucleotide exchange factor activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P258008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124782 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P258008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P258008 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P258008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P258008 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828869 - -
          Affymetrix Rat Gene 1.0 ST Array 10828864 - -
          Affymetrix Rat Gene 1.0 ST Array 10828879 - -
          Affymetrix Rat Gene 1.0 ST Array 10828867 - -
          Affymetrix Rat Gene 1.0 ST Array 10828875 - -
          Affymetrix Rat Gene 1.0 ST Array 10828872 - -
          Affymetrix Rat Gene 1.0 ST Array 10828871 - -
          Affymetrix Rat Gene 1.0 ST Array 10828868 - -
          Affymetrix Rat Gene 1.0 ST Array 10828876 - -
          Affymetrix Rat Gene 1.0 ST Array 10828882 - -
          Affymetrix Rat Gene 1.0 ST Array 10828870 - -
          Affymetrix Rat Gene 1.0 ST Array 10828878 - -
          Affymetrix Rat Gene 1.0 ST Array 10828874 - -
          Affymetrix Rat Gene 1.0 ST Array 10828865 - -
          Affymetrix Rat Gene 1.0 ST Array 10828880 - -
          Affymetrix Rat Gene 1.0 ST Array 10828881 - -
          Affymetrix Rat Gene 1.0 ST Array 10828873 - -
          Affymetrix Rat Gene 1.0 ST Array 10828877 - -
          Affymetrix Rat Gene 1.0 ST Array 10828883 - -
          Affymetrix Rat Gene 1.1 ST Array 10828863 - -
          Rat Genome 230 2.0 Array 1394678_at - -
          Rat Genome 230 2.0 Array 1394678_PM_at - -
          Rat Genome 230 2.0 Array 1397411_at - -
          Rat Genome 230 2.0 Array 1397411_PM_at - -
          Affymetrix HT Rat Focus Array 1397411_at - -
          Affymetrix HT Rat Focus Array 1394678_PM_at - -
          Affymetrix HT Rat Focus Array 1397411_PM_at - -
          Affymetrix HT Rat Focus Array 1394678_at - -
          Affymetrix Rat Clariom S Assay PSR2000002640.rn.1 TC2000000289.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000002642.rn.1 TC2000000289.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000002644.rn.1 TC2000000289.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000002645.rn.1 TC2000000289.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000002643.rn.1 TC2000000289.rn.1 RTA TC
          Rat Expression Set 230 1397411_at - -
          Rat Expression Set 230 1394678_at - -
          Rat Expression Set 230 1397411_PM_at - -
          Rat Expression Set 230 1394678_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828863 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22131718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline