Search
Search
| Gene Symbol: Mpv17l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098240.1 | NP_001091710.1 | 2 - 3 | 435 | 88 | |
| XM_008764515.1 | XP_008762737.1 | 2 - 3 | 5283 | 88 | ||
| XM_008764516.1 | XP_008762738.1 | 2 - 3 | 5282 | 88 | ||
| XM_008764517.1 | XP_008762739.1 | 2 - 3 | 5284 | 88 | ||
| XM_008764518.1 | XP_008762740.1 | 2 - 3 | 424 | 88 | ||
| XM_008764519.1 | XP_008762741.1 | 3 - 4 | 261 | 88 | ||
| GenBank mRNA | BC091193.1 | - | 2 - 3 | 435 | 7373705 | 88 |
Set Membership: |
|