Search
Search
| Gene Symbol: Edar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191899.1 | NP_001178828.1 | 3 - 4 | 360 | 79 | |
| XM_006256381.2 | XP_006256443.1 | 4 - 5 | 653 | 79 | ||
| GenBank mRNA | FQ211923.1 | - | 4 - 5 | 588 | 79 | |
Set Membership: |
|