Search
Search

Trmt61a
LOC103692719| Gene Symbol: Trmt61a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007706.1 | NP_001007707.1 | 2 - 3 | 396 | 77 | |
| XM_008764938.1 | XP_008763160.1 | 1 - 2 | 882 | 77 | ||
| GenBank mRNA | BC079198.1 | - | 2 - 3 | 396 | 7131557 | 77 |
| Gene Symbol: LOC103692719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764975.1 | XP_008763197.1 | 882 | 77 | ||
| XM_008764976.1 | XP_008763198.1 | 395 | 77 | |||
Set Membership: |
|