Search
Search
| Gene Symbol: Akap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114606.1 | NP_001108078.1 | 5 - 6 | 1137 | 68 | |
| XM_006246509.2 | XP_006246571.1 | 5 - 6 | 2734 | 68 | ||
| GenBank mRNA | BC081953.1 | - | 5 - 6 | 1110 | 7122794 | 68 |
Set Membership: |
|