Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01772692_mH
          See other LOC100911937 GE Assays ›
          Gene Symbol:
          LOC100911937 Vom2r26
          Gene Name:
          vomeronasal type-2 receptor 26-like
          vomeronasal 2 receptor, 26
          Gene Aliases:
          -
          Gm1961
          Chromosome Location:
          Chr.3: 17237176 - 17241266 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 63535051 - 63571794 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01772692_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100911937
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749466.1 XP_003749514.1 1 - 2 140 112
          Gene Symbol: Vom2r26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099471.1 NP_001092941.1 5 - 6 1652 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004631.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342576 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143116 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143116 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143116 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P541104 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718502 - -
          Affymetrix Rat Gene 1.0 ST Array 10718499 - -
          Affymetrix Rat Gene 1.0 ST Array 10718498 - -
          Affymetrix Rat Gene 1.0 ST Array 10718503 - -
          Affymetrix Rat Gene 1.0 ST Array 10718500 - -
          Affymetrix Rat Gene 1.0 ST Array 10718501 - -
          Affymetrix Rat Gene 1.1 ST Array 10718497 - -
          Affymetrix Rat Clariom S Assay PSR0100036551.rn.1 TC0100004631.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036554.rn.1 TC0100004631.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036553.rn.1 TC0100004631.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036552.rn.1 TC0100004631.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10718497 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20729273 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651068 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline