Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01772873_g1
          See other SPTA1 GE Assays ›
          Gene Symbol:
          Spta1
          Gene Name:
          spectrin, alpha, erythrocytic 1
          Gene Aliases:
          Spna1
          Chromosome Location:
          Chr.13: 92264231 - 92340091 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01772873_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spta1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011908.3 NP_001011908.2 34 - 35 5051 80
          GenBank mRNA AY238342.1 - 34 - 35 5051 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000822.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spta1

          Gene Ontology Categories:

          Function(s) Process(es)

          lymphocyte homeostasis
          porphyrin-containing compound biosynthetic process
          plasma membrane organization
          biological_process
          regulation of cell shape
          actin cytoskeleton organization
          hemopoiesis
          positive regulation of protein binding
          positive regulation of T cell proliferation
          molecular_function
          calcium ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1300005785.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005791.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005792.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005819.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005790.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005807.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005813.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005816.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005810.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005814.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005788.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005805.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005806.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005820.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005821.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005787.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005789.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005795.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005799.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005800.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005801.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005804.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005808.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005811.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005793.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005796.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005797.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005798.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005802.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005815.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005786.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005794.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005803.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005809.rn.1 TC1300000822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005812.rn.1 TC1300000822.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline