Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01773082_g1
          See other SLC5A10 GE Assays ›
          Gene Symbol:
          Slc5a10
          Gene Name:
          solute carrier family 5 (sodium/sugar cotransporter), member 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 47949777 - 47997803 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01773082_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc5a10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107007.1 NP_001100477.1 14 - 15 1682 72
          XM_008767820.1 XP_008766042.1 14 - 15 1882 72
          GenBank mRNA BC161867.1 - 14 - 15 1875 7107733 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002959.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane transport
          transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P441252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29146 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50445 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P441252 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P441252 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P441252 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P441252 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93979 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58015 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10743461 - -
          Affymetrix Rat Gene 1.0 ST Array 10743463 - -
          Affymetrix Rat Gene 1.0 ST Array 10743453 - -
          Affymetrix Rat Gene 1.0 ST Array 10743458 - -
          Affymetrix Rat Gene 1.0 ST Array 10743459 - -
          Affymetrix Rat Gene 1.0 ST Array 10743466 - -
          Affymetrix Rat Gene 1.0 ST Array 10743456 - -
          Affymetrix Rat Gene 1.0 ST Array 10743457 - -
          Affymetrix Rat Gene 1.0 ST Array 10743455 - -
          Affymetrix Rat Gene 1.0 ST Array 10743465 - -
          Affymetrix Rat Gene 1.0 ST Array 10743464 - -
          Affymetrix Rat Gene 1.0 ST Array 10743460 - -
          Affymetrix Rat Gene 1.0 ST Array 10743467 - -
          Affymetrix Rat Gene 1.0 ST Array 10743462 - -
          Affymetrix Rat Gene 1.0 ST Array 10743454 - -
          Affymetrix Rat Gene 1.1 ST Array 10743452 - -
          Rat Genome 230 2.0 Array 1377196_PM_at - -
          Rat Genome 230 2.0 Array 1377196_at - -
          Affymetrix Rat Clariom S Assay PSR1000026090.rn.1 TC1000002959.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000026092.rn.1 TC1000002959.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10743452 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21876822 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline