Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01773459_g1
          See other PRSS2 GE Assays ›
          Gene Symbol:
          Prss2
          Gene Name:
          protease, serine, 2
          Gene Aliases:
          Ptryss2
          Chromosome Location:
          Chr.4: 70689737 - 70694761 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01773459_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012729.1 NP_036861.1 3 - 4 449 156
          GenBank mRNA V01274.1 - 3 - 4 426 156

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000735.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss2

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          response to nutrient
          digestion
          collagen catabolic process
          serine-type endopeptidase activity
          calcium ion binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104898 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P554904 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70649 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854947 - -
          Affymetrix Rat Gene 1.0 ST Array 10862187 - -
          Affymetrix Rat Gene 1.0 ST Array 10854945 - -
          Affymetrix Rat Gene 1.0 ST Array 10862185 - -
          Affymetrix Rat Gene 1.0 ST Array 10862188 - -
          Affymetrix Rat Gene 1.0 ST Array 10854951 - -
          Affymetrix Rat Gene 1.0 ST Array 10854952 - -
          Affymetrix Rat Gene 1.0 ST Array 10854950 - -
          Affymetrix Rat Gene 1.0 ST Array 10854944 - -
          Affymetrix Rat Gene 1.1 ST Array 10862184 - -
          Affymetrix Rat Gene 1.1 ST Array 10854942 - -
          Affymetrix Rat Gene 1.1 ST Array 10854948 - -
          Rat Genome 230 2.0 Array 1380183_PM_at - -
          Rat Genome 230 2.0 Array 1370126_PM_at - -
          Rat Genome 230 2.0 Array 1380183_at - -
          Rat Genome 230 2.0 Array 1370126_at - -
          Affymetrix HT Rat Focus Array 1370126_PM_at - -
          Affymetrix HT Rat Focus Array 1370126_at - -
          Affymetrix Rat Clariom S Assay PSR0400005405.rn.1 TC0400000735.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005407.rn.1 TC0400000735.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005409.rn.1 TC0400000735.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005408.rn.1 TC0400000735.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005406.rn.1 TC0400000735.rn.1 RTA TC
          Rat Expression Set 230 1370126_PM_at - -
          Rat Expression Set 230 1370126_at - -
          Rat Expression Set 230 1380183_at - -
          Rat Expression Set 230 1380183_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10854942 - -
          Affymetrix Rat Gene 1.0 ST Array 10854948 - -
          Affymetrix Rat Gene 1.0 ST Array 10862184 - -
          Rat Genome U34 Set L00131cds_s_at - -
          Rat Genome U34 Set V01274_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370652 - -
          RatRef-12 Expression BeadChip ILMN_62724 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline