Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01774181_g1
          See other LOC689064 GE Assays ›
          Gene Symbol:
          LOC689064 LOC100134871 LOC103694857
          Gene Name:
          beta-globin
          beta globin minor gene
          hemoglobin subunit beta-2
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.1: 168964202 - 168965568 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 168957460 - 168958836 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906791: 18006 - 19426 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn01774181_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC689064
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001111269.1 NP_001104739.1 129 92
          GenBank mRNA DY319996.1 - 129 92
          FQ209804.1 - 149 92
          FQ209902.1 - 149 92
          FQ210004.1 - 146 92
          FQ210039.1 - 149 92
          FQ210298.1 - 147 92
          FQ210379.1 - 149 92
          FQ210599.1 - 147 92
          FQ210889.1 - 149 92
          FQ211252.1 - 147 92
          FQ211410.1 - 147 92
          FQ211471.1 - 146 92
          FQ211709.1 - 222 92
          FQ215055.1 - 146 92
          FQ217121.1 - 178 92
          FQ217477.1 - 147 92
          FQ218196.1 - 147 92
          FQ218287.1 - 147 92
          FQ218323.1 - 147 92
          FQ221211.1 - 147 92
          FQ225430.1 - 147 92
          FQ225765.1 - 147 92
          FQ226110.1 - 147 92
          FQ230470.1 - 149 92
          FQ230522.1 - 154 92
          FQ230899.1 - 151 92
          Gene Symbol: LOC100134871
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113223.1 NP_001106694.1 1 - 2 146 92
          GenBank mRNA FQ209951.1 - 1 - 2 149 92
          FQ210023.1 - 1 - 2 149 92
          FQ210060.1 - 1 - 2 147 92
          FQ210483.1 - 1 - 2 147 92
          FQ210600.1 - 1 - 2 149 92
          FQ210669.1 - 1 - 2 143 92
          FQ210740.1 - 1 - 2 147 92
          FQ211039.1 - 1 - 2 141 92
          FQ211266.1 - 1 - 2 147 92
          FQ211272.1 - 1 - 2 147 92
          FQ211275.1 - 1 - 2 147 92
          FQ211362.1 - 1 - 2 147 92
          FQ211415.1 - 1 - 2 147 92
          FQ211424.1 - 1 - 2 147 92
          FQ211697.1 - 1 - 2 149 92
          FQ212253.1 - 1 - 2 147 92
          FQ212317.1 - 1 - 2 149 92
          FQ212406.1 - 1 - 2 149 92
          FQ212421.1 - 1 - 2 92
          FQ213080.1 - 1 - 2 176 92
          FQ214057.1 - 1 - 2 147 92
          FQ214129.1 - 1 - 2 147 92
          FQ217673.1 - 1 - 2 147 92
          FQ218002.1 - 1 - 2 148 92
          FQ218191.1 - 1 - 2 147 92
          FQ218246.1 - 1 - 2 119 92
          FQ218266.1 - 1 - 2 185 92
          FQ218689.1 - 1 - 2 147 92
          FQ218821.1 - 1 - 2 147 92
          FQ219133.1 - 1 - 2 178 92
          FQ221588.1 - 1 - 2 145 92
          FQ222844.1 - 1 - 2 147 92
          FQ224149.1 - 1 - 2 147 92
          FQ224301.1 - 1 - 2 147 92
          FQ224598.1 - 1 - 2 147 92
          FQ224632.1 - 1 - 2 147 92
          FQ224738.1 - 1 - 2 147 92
          FQ224763.1 - 1 - 2 186 92
          FQ225377.1 - 1 - 2 147 92
          FQ225739.1 - 1 - 2 149 92
          FQ226460.1 - 1 - 2 147 92
          FQ226535.1 - 1 - 2 147 92
          FQ226609.1 - 1 - 2 149 92
          FQ226955.1 - 1 - 2 147 92
          FQ228555.1 - 1 - 2 147 92
          FQ230881.1 - 1 - 2 149 92
          FQ232298.1 - 291 92
          FQ232442.1 - 1 - 2 178 92
          FQ233001.1 - 1 - 2 147 92
          FQ234495.1 - 1 - 2 148 92
          X05080.1 - 1 - 2 146 92
          X16418.1 - 1 - 2 112 92
          Gene Symbol: LOC103694857
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008773801.1 XP_008772023.1 1 - 2 183 92

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          oxygen transport
          positive regulation of cell death
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          protein heterooligomerization
          oxidation-reduction process
          renal absorption
          platelet aggregation
          oxygen transporter activity
          iron ion binding
          oxygen binding
          heme binding
          peroxidase activity
          hemoglobin binding
          haptoglobin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724313 - -
          Affymetrix Rat Gene 1.0 ST Array 10724314 - -
          Affymetrix Rat Gene 1.0 ST Array 10724312 - -
          Affymetrix Rat Gene 1.1 ST Array 10724311 - -
          Rat Genome 230 2.0 Array 1371245_PM_a_at - -
          Rat Genome 230 2.0 Array 1371245_a_at - -
          Rat Genome 230 2.0 Array 1371102_PM_x_at - -
          Rat Genome 230 2.0 Array 1371102_x_at - -
          Affymetrix HT Rat Focus Array 1371102_x_at - -
          Affymetrix HT Rat Focus Array 1371102_PM_x_at - -
          Affymetrix Rat Clariom S Assay PSR0100017230.rn.1 TC0100002239.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100017232.rn.1 TC0100002239.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100017231.rn.1 TC0100002239.rn.1 RTA TC
          Rat Expression Set 230 1371245_a_at - -
          Rat Expression Set 230 1371102_PM_x_at - -
          Rat Expression Set 230 1371102_x_at - -
          Rat Expression Set 230 1371245_PM_a_at - -
          Rat Genome U34 Set rc_AI179576_s_at - -
          Rat Genome U34 Set rc_AI012182_s_at - -
          Rat Genome U34 Set rc_AI011943_at - -
          Rat Genome U34 Set rc_AI177992_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline