Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01774390_g1
          See other USP12 GE Assays ›
          Gene Symbol:
          Usp12
          Gene Name:
          ubiquitin specific peptidase 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 10037516 - 10090545 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01774390_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Usp12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166576.1 NP_001160048.1 8 - 9 1011 167
          GenBank mRNA FN557299.1 - 8 - 9 1011 167

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          biological_process
          protein deubiquitination
          molecular_function
          cysteine-type endopeptidase activity
          thiol-dependent ubiquitin-specific protease activity
          cysteine-type peptidase activity
          thiol-dependent ubiquitinyl hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094831 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P555884 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094831 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P555884 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094831 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P555884 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P555884 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.1 ST Array 10756500 - -
          Affymetrix Rat Gene 1.1 ST Array 10756510 - -
          Affymetrix Rat Gene 1.1 ST Array 10893327 - -
          Affymetrix Rat Gene 1.1 ST Array 10940058 - -
          Rat Genome 230 2.0 Array 1385035_at - -
          Rat Genome 230 2.0 Array 1385035_PM_at - -
          Affymetrix HT Rat Focus Array 1385035_at - -
          Affymetrix HT Rat Focus Array 1385035_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200001184.rn.1 TC1200000169.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001185.rn.1 TC1200000169.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001186.rn.1 TC1200000169.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001188.rn.1 TC1200000169.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001187.rn.1 TC1200000169.rn.1 RTA TC
          Rat Expression Set 230 1385035_at - -
          Rat Expression Set 230 1385035_PM_at - -
          Rat Genome U34 Set rc_AI639241_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline