Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01775763_g1
          See other GAPDH GE Assays ›
          Gene Symbol:
          Gapdh
          Gene Name:
          glyceraldehyde-3-phosphate dehydrogenase
          Gene Aliases:
          BARS-38, Gapd
          Chromosome Location:
          Chr.4: 157676396 - 157680271 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01775763_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gapdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017008.4 NP_058704.1 8 1153 174
          GenBank mRNA AB017801.1 - 8 1119 174
          BC059110.1 - 8 1154 6919611 174
          BC087743.1 - 8 1122 7321006 174
          M11561.1 - 2 280 174
          M17701.1 - 7 175
          M29341.1 - 3 376 174
          X02231.1 - 8 1149 174

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gapdh

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          carbohydrate metabolic process
          gluconeogenesis
          glycolytic process
          regulation of translation
          apoptotic process
          multicellular organism development
          negative regulation of translation
          peptidyl-cysteine S-trans-nitrosylation
          protein stabilization
          neuron apoptotic process
          oxidation-reduction process
          response to ammonium ion
          cellular response to interferon-gamma
          glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
          protein binding
          microtubule binding
          peptidyl-cysteine S-nitrosylase activity
          identical protein binding
          NADP binding
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P232432 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052510 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18824 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P232432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003046 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P232432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003046 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P232432 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052510 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003046 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P363377 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P193181 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700919988 - -
          Agilent Rat cDNA Microarray (G4105A) 700919988 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782756 - -
          Affymetrix Rat Gene 1.0 ST Array 10787810 - -
          Affymetrix Rat Gene 1.0 ST Array 10787809 - -
          Affymetrix Rat Gene 1.0 ST Array 10865629 - -
          Affymetrix Rat Gene 1.0 ST Array 10865623 - -
          Affymetrix Rat Gene 1.0 ST Array 10714737 - -
          Affymetrix Rat Gene 1.0 ST Array 10916015 - -
          Affymetrix Rat Gene 1.0 ST Array 10754992 - -
          Affymetrix Rat Gene 1.0 ST Array 10813722 - -
          Affymetrix Rat Gene 1.0 ST Array 10759493 - -
          Affymetrix Rat Gene 1.0 ST Array 10937401 - -
          Affymetrix Rat Gene 1.0 ST Array 10840273 - -
          Affymetrix Rat Gene 1.0 ST Array 10806312 - -
          Affymetrix Rat Gene 1.0 ST Array 10934568 - -
          Affymetrix Rat Gene 1.0 ST Array 10865624 - -
          Affymetrix Rat Gene 1.0 ST Array 10799941 - -
          Affymetrix Rat Gene 1.0 ST Array 10864403 - -
          Affymetrix Rat Gene 1.0 ST Array 10865628 - -
          Affymetrix Rat Gene 1.0 ST Array 10769886 - -
          Affymetrix Rat Gene 1.0 ST Array 10905816 - -
          Affymetrix Rat Gene 1.0 ST Array 10865625 - -
          Affymetrix Rat Gene 1.0 ST Array 10919556 - -
          Affymetrix Rat Gene 1.0 ST Array 10787811 - -
          Affymetrix Rat Gene 1.0 ST Array 10754881 - -
          Affymetrix Rat Gene 1.0 ST Array 10776680 - -
          Affymetrix Rat Gene 1.0 ST Array 10787302 - -
          Affymetrix Rat Gene 1.0 ST Array 10870019 - -
          Affymetrix Rat Gene 1.0 ST Array 10909369 - -
          Affymetrix Rat Gene 1.0 ST Array 10748801 - -
          Affymetrix Rat Gene 1.0 ST Array 10888367 - -
          Affymetrix Rat Gene 1.0 ST Array 10865626 - -
          Affymetrix Rat Gene 1.0 ST Array 10909370 - -
          Affymetrix Rat Gene 1.0 ST Array 10870016 - -
          Affymetrix Rat Gene 1.0 ST Array 10841483 - -
          Affymetrix Rat Gene 1.0 ST Array 10935264 - -
          Affymetrix Rat Gene 1.0 ST Array 10870017 - -
          Affymetrix Rat Gene 1.0 ST Array 10868254 - -
          Affymetrix Rat Gene 1.0 ST Array 10787070 - -
          Affymetrix Rat Gene 1.0 ST Array 10905818 - -
          Affymetrix Rat Gene 1.0 ST Array 10870015 - -
          Affymetrix Rat Gene 1.0 ST Array 10870018 - -
          Affymetrix Rat Gene 1.0 ST Array 10754882 - -
          Affymetrix Rat Gene 1.0 ST Array 10778889 - -
          Affymetrix Rat Gene 1.0 ST Array 10841482 - -
          Affymetrix Rat Gene 1.0 ST Array 10806313 - -
          Affymetrix Rat Gene 1.0 ST Array 10876835 - -
          Affymetrix Rat Gene 1.0 ST Array 10770542 - -
          Affymetrix Rat Gene 1.0 ST Array 10751600 - -
          Affymetrix Rat Gene 1.0 ST Array 10759494 - -
          Affymetrix Rat Gene 1.0 ST Array 10865627 - -
          Affymetrix Rat Gene 1.0 ST Array 10868253 - -
          Affymetrix Rat Gene 1.0 ST Array 10802514 - -
          Affymetrix Rat Gene 1.0 ST Array 10937402 - -
          Affymetrix Rat Gene 1.0 ST Array 10934567 - -
          Affymetrix Rat Gene 1.1 ST Array 10888366 - -
          Affymetrix Rat Gene 1.1 ST Array 10905817 - -
          Affymetrix Rat Gene 1.1 ST Array 10865622 - -
          Affymetrix Rat Gene 1.1 ST Array 10934566 - -
          Affymetrix Rat Gene 1.1 ST Array 10802513 - -
          Affymetrix Rat Gene 1.1 ST Array 10937400 - -
          Affymetrix Rat Gene 1.1 ST Array 10876834 - -
          Affymetrix Rat Gene 1.1 ST Array 10813721 - -
          Affymetrix Rat Gene 1.1 ST Array 10787301 - -
          Affymetrix Rat Gene 1.1 ST Array 10919555 - -
          Affymetrix Rat Gene 1.1 ST Array 10778888 - -
          Affymetrix Rat Gene 1.1 ST Array 10759492 - -
          Affymetrix Rat Gene 1.1 ST Array 10868252 - -
          Affymetrix Rat Gene 1.1 ST Array 10806311 - -
          Affymetrix Rat Gene 1.1 ST Array 10935263 - -
          Affymetrix Rat Gene 1.1 ST Array 10770541 - -
          Affymetrix Rat Gene 1.1 ST Array 10769885 - -
          Affymetrix Rat Gene 1.1 ST Array 10748800 - -
          Affymetrix Rat Gene 1.1 ST Array 10864402 - -
          Affymetrix Rat Gene 1.1 ST Array 10799940 - -
          Affymetrix Rat Gene 1.1 ST Array 10751599 - -
          Affymetrix Rat Gene 1.1 ST Array 10787069 - -
          Affymetrix Rat Gene 1.1 ST Array 10905815 - -
          Affymetrix Rat Gene 1.1 ST Array 10754880 - -
          Affymetrix Rat Gene 1.1 ST Array 10776679 - -
          Affymetrix Rat Gene 1.1 ST Array 10714736 - -
          Affymetrix Rat Gene 1.1 ST Array 10787808 - -
          Affymetrix Rat Gene 1.1 ST Array 10870014 - -
          Affymetrix Rat Gene 1.1 ST Array 10782755 - -
          Affymetrix Rat Gene 1.1 ST Array 10754991 - -
          Affymetrix Rat Gene 1.1 ST Array 10916014 - -
          Affymetrix Rat Gene 1.1 ST Array 10909368 - -
          Affymetrix Rat Gene 1.1 ST Array 10841481 - -
          Affymetrix Rat Gene 1.1 ST Array 10840272 - -
          Rat Genome 230 2.0 Array AFFX_Rat_GAPDH_5_at - -
          Rat Genome 230 2.0 Array AFFX_Rat_GAPDH_M_at - -
          Rat Genome 230 2.0 Array 1367557_s_at - -
          Rat Genome 230 2.0 Array AFFX_Rat_GAPDH_3_at - -
          Rat Genome 230 2.0 Array 1367557_PM_s_at - -
          Affymetrix HT Rat Focus Array 1367557_s_at - -
          Affymetrix HT Rat Focus Array 1367557_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0400026683.rn.1 TC0400003755.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026682.rn.1 TC0400003755.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026685.rn.1 TC0400003755.rn.1 RTA TC
          Rat Expression Set 230 AFFX_Rat_GAPDH_3_at - -
          Rat Expression Set 230 AFFX_Rat_GAPDH_5_at - -
          Rat Expression Set 230 1367557_s_at - -
          Rat Expression Set 230 AFFX_Rat_GAPDH_M_at - -
          Rat Expression Set 230 1367557_PM_s_at - -
          Rat Expression Set 230 AFFX_Rat_GAPDH_3_at - -
          Rat Expression Set 230 AFFX_Rat_GAPDH_5_at - -
          Rat Expression Set 230 AFFX_Rat_GAPDH_M_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864402 - -
          Affymetrix Rat Gene 1.0 ST Array 10754880 - -
          Affymetrix Rat Gene 1.0 ST Array 10840272 - -
          Affymetrix Rat Gene 1.0 ST Array 10870014 - -
          Affymetrix Rat Gene 1.0 ST Array 10905815 - -
          Affymetrix Rat Gene 1.0 ST Array 10876834 - -
          Affymetrix Rat Gene 1.0 ST Array 10909368 - -
          Affymetrix Rat Gene 1.0 ST Array 10865622 - -
          Affymetrix Rat Gene 1.0 ST Array 10905817 - -
          Affymetrix Rat Gene 1.0 ST Array 10787808 - -
          Affymetrix Rat Gene 1.0 ST Array 10802513 - -
          Affymetrix Rat Gene 1.0 ST Array 10919555 - -
          Affymetrix Rat Gene 1.0 ST Array 10748800 - -
          Affymetrix Rat Gene 1.0 ST Array 10769885 - -
          Affymetrix Rat Gene 1.0 ST Array 10868252 - -
          Affymetrix Rat Gene 1.0 ST Array 10776679 - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_at - -
          Rat Genome U34 Set M17701_s_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_5_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_st - -
          Rat Genome U34 Set M17701_s_at - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_M_st - -
          Rat Genome U34 Set AFFX_Rat_GAPDH_3_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21551650 - -
          ABI Rat Genome Survey Microarray 21826176 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649859 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline