Search
Search

St3gal4| Gene Symbol: St3gal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203337.2 | NP_976082.1 | 4 - 5 | 421 | 75 | |
| XM_006242789.2 | XP_006242851.1 | 5 - 6 | 537 | 75 | ||
| XM_008766058.1 | XP_008764280.1 | 4 - 5 | 923 | 75 | ||
| XM_008766059.1 | XP_008764281.1 | 4 - 5 | 912 | 75 | ||
| XM_008766060.1 | XP_008764282.1 | 4 - 5 | 1371 | 75 | ||
| GenBank mRNA | AJ626825.1 | - | 3 - 4 | 185 | 75 | |
| BC089057.1 | - | 4 - 5 | 421 | 7315905 | 75 | |
Set Membership: |
|