Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01789798_s1
          See other HBA1 GE Assays ›
          Gene Symbol:
          Hba1 Hbb
          Gene Name:
          hemoglobin, alpha 1
          hemoglobin, beta
          Gene Aliases:
          HBAM, Hba-a1, Hba-a2
          -
          Chromosome Location:
          Chr.10: 15602794 - 15603649 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 168971269 - 168972680 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01789798_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hba1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013096.1 NP_037228.1 3 498 102
          GenBank mRNA BC059150.1 - 3 497 6918464 102
          BC160818.1 - 3 481 7375173 102
          FQ209733.1 - 3 498 102
          FQ209744.1 - 3 498 102
          FQ209909.1 - 3 498 102
          FQ209912.1 - 3 498 102
          FQ210001.1 - 3 498 102
          FQ210150.1 - 3 496 102
          FQ210186.1 - 3 496 102
          FQ210277.1 - 3 496 102
          FQ210349.1 - 3 496 102
          FQ210690.1 - 3 497 102
          FQ210723.1 - 3 496 102
          FQ210862.1 - 3 496 102
          FQ210866.1 - 3 496 102
          FQ211077.1 - 3 498 102
          FQ211217.1 - 3 496 102
          FQ211232.1 - 3 497 102
          FQ211239.1 - 3 496 102
          FQ211351.1 - 3 496 102
          FQ211374.1 - 3 496 102
          FQ211431.1 - 3 496 102
          FQ211513.1 - 3 496 102
          FQ211520.1 - 3 496 102
          FQ211755.1 - 3 496 102
          FQ211911.1 - 3 496 102
          FQ211972.1 - 3 496 102
          FQ212025.1 - 3 498 102
          FQ212144.1 - 3 498 102
          FQ212153.1 - 3 498 102
          FQ212186.1 - 3 496 102
          FQ212232.1 - 3 498 102
          FQ212457.1 - 3 496 102
          FQ212854.1 - 3 496 102
          FQ213301.1 - 3 102
          FQ214066.1 - 3 496 102
          FQ214223.1 - 3 496 102
          FQ216243.1 - 3 496 102
          FQ217723.1 - 3 496 102
          FQ217724.1 - 3 495 102
          FQ217869.1 - 3 496 102
          FQ217988.1 - 3 498 102
          FQ218117.1 - 3 496 102
          FQ218146.1 - 496 102
          FQ218257.1 - 3 552 102
          FQ218612.1 - 3 495 102
          FQ218968.1 - 3 498 102
          FQ219715.1 - 3 496 102
          FQ220786.1 - 3 496 102
          FQ221656.1 - 3 497 102
          FQ221912.1 - 3 498 102
          FQ221952.1 - 3 496 102
          FQ222672.1 - 3 496 102
          FQ222750.1 - 3 498 102
          FQ223017.1 - 3 497 102
          FQ224581.1 - 3 496 102
          FQ225427.1 - 3 497 102
          FQ225440.1 - 3 497 102
          FQ225777.1 - 3 496 102
          FQ225808.1 - 3 499 102
          FQ226118.1 - 3 498 102
          FQ226175.1 - 3 102
          FQ226203.1 - 3 496 102
          FQ226615.1 - 3 496 102
          FQ227082.1 - 3 496 102
          FQ228301.1 - 3 496 102
          FQ228436.1 - 496 102
          FQ230483.1 - 3 498 102
          FQ230901.1 - 3 496 102
          FQ231201.1 - 3 488 102
          FQ231290.1 - 3 496 102
          FQ232272.1 - 3 496 102
          FQ232481.1 - 3 496 102
          M17083.1 - 3 498 102
          Gene Symbol: Hbb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA FQ209822.1 - 832 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004401.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          oxygen transport
          negative regulation of blood pressure
          protein heterooligomerization
          regulation of sensory perception of pain
          beta-amyloid binding
          oxygen transporter activity
          iron ion binding
          protein binding
          oxygen binding
          heme binding
          organic acid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032502 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032499 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032502 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032499 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032502 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032499 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032499 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032502 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700293050 - -
          Agilent Rat cDNA Microarray (G4105A) 700293050 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741779 - -
          Affymetrix Rat Gene 1.0 ST Array 10741780 - -
          Affymetrix Rat Gene 1.0 ST Array 10741760 - -
          Affymetrix Rat Gene 1.0 ST Array 10741781 - -
          Affymetrix Rat Gene 1.0 ST Array 10741759 - -
          Affymetrix Rat Gene 1.0 ST Array 10741758 - -
          Affymetrix Rat Gene 1.0 ST Array 10741757 - -
          Affymetrix Rat Gene 1.1 ST Array 10741756 - -
          Affymetrix Rat Gene 1.1 ST Array 10741778 - -
          Rat Genome 230 2.0 Array 1370239_at - -
          Rat Genome 230 2.0 Array 1388608_x_at - -
          Rat Genome 230 2.0 Array 1370240_x_at - -
          Rat Genome 230 2.0 Array 1370239_PM_at - -
          Rat Genome 230 2.0 Array 1370240_PM_x_at - -
          Rat Genome 230 2.0 Array 1375519_PM_at - -
          Rat Genome 230 2.0 Array 1375519_at - -
          Rat Genome 230 2.0 Array 1388608_PM_x_at - -
          Affymetrix Rat Clariom S Assay PSR1000041321.rn.1 TC1000004401.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041325.rn.1 TC1000004401.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041322.rn.1 TC1000004401.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041326.rn.1 TC1000004401.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041323.rn.1 TC1000004401.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041319.rn.1 TC1000004401.rn.1 RTA TC
          Rat Expression Set 230 1388608_PM_x_at - -
          Rat Expression Set 230 1388608_x_at - -
          Rat Expression Set 230 1370239_PM_at - -
          Rat Expression Set 230 1370239_at - -
          Rat Expression Set 230 1370240_x_at - -
          Rat Expression Set 230 1370240_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741778 - -
          Affymetrix Rat Gene 1.0 ST Array 10741756 - -
          Rat Genome U34 Set X56325mRNA_s_at - -
          Rat Genome U34 Set rc_AI012230_s_at - -
          Rat Genome U34 Set rc_AI013718_s_at - -
          Rat Genome U34 Set rc_AI013718_i_at - -
          Rat Genome U34 Set rc_AA851497_f_at - -
          Rat Genome U34 Set rc_AI230970_f_at - -
          Rat Genome U34 Set rc_AI236360_f_at - -
          Rat Genome U34 Set rc_AI179971_f_at - -
          Rat Genome U34 Set rc_AI179971_i_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356639 - -
          RatRef-12 Expression BeadChip ILMN_65102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline