Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02052476_s1
          See other VOM1R90 GE Assays ›
          Gene Symbol:
          Vom1r90
          Gene Name:
          vomeronasal 1 receptor 90
          Gene Aliases:
          V1ra16
          Chromosome Location:
          Chr.4: 121675261 - 121684264 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02052476_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vom1r90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153729.2 NP_714951.1 3 1237 111
          GenBank mRNA U36898.1 - 3 1257 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vom1r90

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804322 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171452 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069759 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069759 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88081 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57452 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864006 - -
          Affymetrix Rat Gene 1.0 ST Array 10864005 - -
          Affymetrix Rat Gene 1.0 ST Array 10864002 - -
          Affymetrix Rat Gene 1.0 ST Array 10864004 - -
          Affymetrix Rat Gene 1.0 ST Array 10864003 - -
          Affymetrix Rat Gene 1.1 ST Array 10864001 - -
          Rat Genome 230 2.0 Array 1370720_at - -
          Rat Genome 230 2.0 Array 1370720_PM_at - -
          Affymetrix HT Rat Focus Array 1370720_at - -
          Affymetrix HT Rat Focus Array 1370720_PM_at - -
          Rat Expression Set 230 1370720_at - -
          Rat Expression Set 230 1370720_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864001 - -
          Rat Genome U34 Set U36898_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21728635 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351192 - -
          RatRef-12 Expression BeadChip ILMN_63690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline