Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02070661_s1
          See other P2RY2 GE Assays ›
          Gene Symbol:
          P2ry2
          Gene Name:
          purinergic receptor P2Y, G-protein coupled, 2
          Gene Aliases:
          P2Y2
          Chromosome Location:
          Chr.1: 166031228 - 166045423 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02070661_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2ry2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017255.1 NP_058951.1 3 1840 137
          XM_006229757.2 XP_006229819.1 3 1623 137
          GenBank mRNA BC061754.1 - 3 1843 5599916 137
          L46865.1 - 3 1840 137
          U56839.1 - 2 1498 137

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2ry2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          aging
          positive regulation of neuron projection development
          positive regulation of smooth muscle cell migration
          sensory perception of pain
          negative regulation of ossification
          positive regulation of prostaglandin secretion
          positive regulation of chemokine production
          G-protein coupled purinergic nucleotide receptor signaling pathway
          wound healing
          regulation of vasodilation
          positive regulation of penile erection
          positive regulation of mucus secretion
          response to growth factor
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to purine-containing compound
          protein binding
          ATP binding
          A1 adenosine receptor binding
          G-protein coupled purinergic nucleotide receptor activity
          UTP-activated nucleotide receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P520159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62357 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P520159 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P520159 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P520159 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P520159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91965 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724045 - -
          Affymetrix Rat Gene 1.0 ST Array 10724044 - -
          Affymetrix Rat Gene 1.0 ST Array 10724043 - -
          Affymetrix Rat Gene 1.0 ST Array 10724046 - -
          Affymetrix Rat Gene 1.1 ST Array 10724042 - -
          Rat Genome 230 2.0 Array 1368940_PM_at - -
          Rat Genome 230 2.0 Array 1368940_at - -
          Affymetrix HT Rat Focus Array 1368940_PM_at - -
          Affymetrix HT Rat Focus Array 1368940_at - -
          Affymetrix Rat Clariom S Assay PSR0100048891.rn.1 TC0100006150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100048888.rn.1 TC0100006150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100048889.rn.1 TC0100006150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100048885.rn.1 TC0100006150.rn.1 RTA TC
          Rat Expression Set 230 1368940_at - -
          Rat Expression Set 230 1368940_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724042 - -
          Rat Genome U34 Set U56839_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20951074 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364530 - -
          RatRef-12 Expression BeadChip ILMN_61673 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline