Search
Search

Edaradd| Gene Symbol: Edaradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771925.1 | XP_008770147.1 | 5 | 1956 | 152 | |
| XM_008771926.1 | XP_008770148.1 | 7 | 1996 | 152 | ||
| XM_574055.6 | XP_574055.4 | 6 | 2108 | 152 | ||
Set Membership: |
|