Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02104120_s1
          See other RGD1560212 GE Assays ›
          Gene Symbol:
          RGD1560212 LOC102547036
          Gene Name:
          similar to DNA segment, Chr 18, Wayne State University 98, expressed
          uncharacterized LOC102547036
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.18: 76725229 - 76731645 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 76731395 - 76737262 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn02104120_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1560212
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077676.1 NP_001071144.1 4 1512 160
          XM_006254957.2 XP_006255019.1 3 1467 160
          XM_008772164.1 XP_008770386.1 4 1488 160
          GenBank mRNA BC127540.1 - 4 1512 7301991 160
          FQ234858.1 - 3 1477 160
          Gene Symbol: LOC102547036
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008772213.1 XP_008770435.1 2 160
          XM_008772214.1 XP_008770436.1 2 160

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          acetyltransferase

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P212424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502032 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P212424 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P212424 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P212424 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P212424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83216 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802950 - -
          Affymetrix Rat Gene 1.0 ST Array 10802953 - -
          Affymetrix Rat Gene 1.0 ST Array 10802952 - -
          Affymetrix Rat Gene 1.0 ST Array 10802951 - -
          Affymetrix Rat Gene 1.1 ST Array 10802949 - -
          Rat Genome 230 2.0 Array 1371971_at - -
          Rat Genome 230 2.0 Array 1367493_at - -
          Rat Genome 230 2.0 Array 1371971_PM_at - -
          Rat Genome 230 2.0 Array 1367493_PM_at - -
          Affymetrix HT Rat Focus Array 1367493_PM_at - -
          Affymetrix HT Rat Focus Array 1371971_PM_at - -
          Affymetrix HT Rat Focus Array 1371971_at - -
          Affymetrix HT Rat Focus Array 1367493_at - -
          Rat Expression Set 230 1371971_PM_at - -
          Rat Expression Set 230 1367493_PM_at - -
          Rat Expression Set 230 1371971_at - -
          Rat Expression Set 230 1367493_at - -
          Rat Expression Set 230 1367493_at - -
          Rat Expression Set 230 1367493_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802949 - -
          Rat Genome U34 Set rc_AI169269_at - -
          Rat Genome U34 Set rc_AI231077_at - -
          Rat Genome U34 Set rc_AI179355_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21153234 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370745 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline