Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02108832_s1
          See other PIWIL1 GE Assays ›
          Gene Symbol:
          Piwil1
          Gene Name:
          piwi-like RNA-mediated gene silencing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 31609053 - 31629881 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02108832_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Piwil1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108853.1 NP_001102323.1 23 3956 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          spermatid development
          piRNA metabolic process
          single-stranded RNA binding
          mRNA binding
          protein kinase binding
          piRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P555309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907193 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48089 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P555309 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P555309 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P555309 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555309 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761483 - -
          Affymetrix Rat Gene 1.0 ST Array 10761480 - -
          Affymetrix Rat Gene 1.0 ST Array 10761475 - -
          Affymetrix Rat Gene 1.0 ST Array 10761476 - -
          Affymetrix Rat Gene 1.0 ST Array 10761472 - -
          Affymetrix Rat Gene 1.0 ST Array 10761485 - -
          Affymetrix Rat Gene 1.0 ST Array 10761477 - -
          Affymetrix Rat Gene 1.0 ST Array 10761486 - -
          Affymetrix Rat Gene 1.0 ST Array 10761484 - -
          Affymetrix Rat Gene 1.0 ST Array 10761481 - -
          Affymetrix Rat Gene 1.0 ST Array 10761487 - -
          Affymetrix Rat Gene 1.0 ST Array 10761482 - -
          Affymetrix Rat Gene 1.0 ST Array 10761473 - -
          Affymetrix Rat Gene 1.0 ST Array 10761478 - -
          Affymetrix Rat Gene 1.0 ST Array 10761490 - -
          Affymetrix Rat Gene 1.0 ST Array 10761489 - -
          Affymetrix Rat Gene 1.0 ST Array 10761474 - -
          Affymetrix Rat Gene 1.0 ST Array 10761479 - -
          Affymetrix Rat Gene 1.0 ST Array 10761488 - -
          Affymetrix Rat Gene 1.0 ST Array 10761471 - -
          Affymetrix Rat Gene 1.1 ST Array 10761470 - -
          Affymetrix Rat Clariom S Assay PSR1200012434.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012418.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012423.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012425.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012435.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012420.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012424.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012426.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012427.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012436.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012412.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012430.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012431.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012417.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012410.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012421.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012429.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012411.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012416.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012419.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012422.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012428.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012432.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012433.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012437.rn.1 TC1200001466.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10761470 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21053114 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362153 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline