Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02111291_s1
          See other BIN2A GE Assays ›
          Gene Symbol:
          Bin2a
          Gene Name:
          beta-galactosidase-like protein
          Gene Aliases:
          Glb1l4
          Chromosome Location:
          Chr.8: 27935385 - 27974006 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02111291_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bin2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009524.1 NP_001009524.1 21 2398 96
          XM_006242737.2 XP_006242799.1 21 2276 96
          GenBank mRNA AF414431.1 - 21 2252 96
          AY040870.1 - 21 2398 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bin2a

          Panther Classification:

          Molecular Function -

          galactosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          beta-galactosidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003888 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003888 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003888 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P114138 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103761 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8765 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15975 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915908 - -
          Affymetrix Rat Gene 1.0 ST Array 10915890 - -
          Affymetrix Rat Gene 1.0 ST Array 10915893 - -
          Affymetrix Rat Gene 1.0 ST Array 10915909 - -
          Affymetrix Rat Gene 1.0 ST Array 10915895 - -
          Affymetrix Rat Gene 1.0 ST Array 10915900 - -
          Affymetrix Rat Gene 1.0 ST Array 10915888 - -
          Affymetrix Rat Gene 1.0 ST Array 10915897 - -
          Affymetrix Rat Gene 1.0 ST Array 10915892 - -
          Affymetrix Rat Gene 1.0 ST Array 10915905 - -
          Affymetrix Rat Gene 1.0 ST Array 10915903 - -
          Affymetrix Rat Gene 1.0 ST Array 10915906 - -
          Affymetrix Rat Gene 1.0 ST Array 10915896 - -
          Affymetrix Rat Gene 1.0 ST Array 10915904 - -
          Affymetrix Rat Gene 1.0 ST Array 10915898 - -
          Affymetrix Rat Gene 1.0 ST Array 10915899 - -
          Affymetrix Rat Gene 1.0 ST Array 10915907 - -
          Affymetrix Rat Gene 1.0 ST Array 10915894 - -
          Affymetrix Rat Gene 1.0 ST Array 10915889 - -
          Affymetrix Rat Gene 1.0 ST Array 10915902 - -
          Affymetrix Rat Gene 1.0 ST Array 10915901 - -
          Affymetrix Rat Gene 1.0 ST Array 10915891 - -
          Affymetrix Rat Gene 1.1 ST Array 10915887 - -
          Rat Genome 230 2.0 Array 1382669_PM_at - -
          Rat Genome 230 2.0 Array 1382669_at - -
          Affymetrix HT Rat Focus Array 1382669_PM_at - -
          Affymetrix HT Rat Focus Array 1382669_at - -
          Affymetrix Rat Clariom S Assay PSR0800017409.rn.1 TC0800002138.rn.1 RTA TC
          Rat Expression Set 230 1382669_at - -
          Rat Expression Set 230 1382669_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10915887 - -
          Rat Genome U34 Set rc_AI236759_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21442631 - -
          ABI Rat Genome Survey Microarray 21792200 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360965 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline