Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02113326_s1
          See other RGD1565222 GE Assays ›
          Gene Symbol:
          RGD1565222
          Gene Name:
          similar to RIKEN cDNA 4931414P19
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 33225172 - 33259916 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02113326_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1565222
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109101.1 NP_001102571.1 7 2442 164
          XM_006252004.2 XP_006252066.1 7 2740 164
          XM_006252005.2 XP_006252067.1 7 2398 164
          XM_006252006.2 XP_006252068.1 2358 164

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144247 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065492 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065492 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065492 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783597 - -
          Affymetrix Rat Gene 1.0 ST Array 10783598 - -
          Affymetrix Rat Gene 1.0 ST Array 10783596 - -
          Affymetrix Rat Gene 1.0 ST Array 10783594 - -
          Affymetrix Rat Gene 1.0 ST Array 10783595 - -
          Affymetrix Rat Gene 1.0 ST Array 10783593 - -
          Affymetrix Rat Gene 1.1 ST Array 10783592 - -
          Rat Genome 230 2.0 Array 1374937_PM_at - -
          Rat Genome 230 2.0 Array 1374937_at - -
          Affymetrix HT Rat Focus Array 1374937_at - -
          Affymetrix HT Rat Focus Array 1374937_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500010595.rn.1 TC1500001673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010594.rn.1 TC1500001673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010597.rn.1 TC1500001673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010598.rn.1 TC1500001673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010599.rn.1 TC1500001673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010596.rn.1 TC1500001673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010602.rn.1 TC1500001673.rn.1 RTA TC
          Rat Expression Set 230 1374937_PM_at - -
          Rat Expression Set 230 1374937_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783592 - -
          Rat Genome U34 Set rc_AI070891_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20961594 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349409 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline