Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02114291_s1
          See other RPL37A GE Assays ›
          Gene Symbol:
          Rpl37a
          Gene Name:
          ribosomal protein L37a
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02114291_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl37a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108801.1 NP_001102271.1 3 535 93
          GenBank mRNA CF108195.1 - 2 484 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Endogenous Controls Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Endogenous Controls 1700 Inventoried Rat_Endogenous_Controls Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1008356 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015051 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1008356 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015051 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123277 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123277 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015051 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1008356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924218 - -
          Affymetrix Rat Gene 1.0 ST Array 10879917 - -
          Affymetrix Rat Gene 1.0 ST Array 10924219 - -
          Affymetrix Rat Gene 1.0 ST Array 10879918 - -
          Affymetrix Rat Gene 1.0 ST Array 10731731 - -
          Affymetrix Rat Gene 1.0 ST Array 10731730 - -
          Affymetrix Rat Gene 1.0 ST Array 10924220 - -
          Affymetrix Rat Gene 1.1 ST Array 10879915 - -
          Affymetrix Rat Gene 1.1 ST Array 10924217 - -
          Affymetrix Rat Gene 1.1 ST Array 10731729 - -
          Affymetrix Rat Gene 1.0 ST Array 10924217 - -
          Rat Genome U34 Set rc_AA900050_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21503307 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline