Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02132493_s1
          See other IRS1 GE Assays ›
          Gene Symbol:
          Irs1
          Gene Name:
          insulin receptor substrate 1
          Gene Aliases:
          IRS1IRM
          Chromosome Location:
          Chr.9: 88033668 - 88086488 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02132493_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Irs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012969.1 NP_037101.1 2 5228 147
          GenBank mRNA X58375.1 - 2 5228 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Irs1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of mesenchymal cell proliferation
          insulin receptor signaling pathway
          regulation of gene expression
          positive regulation of glucose metabolic process
          phosphatidylinositol 3-kinase signaling
          lipid catabolic process
          insulin secretion
          positive regulation of cell migration
          mammary gland development
          response to caffeine
          positive regulation of fatty acid beta-oxidation
          response to insulin
          cellular response to insulin stimulus
          protein localization to nucleus
          positive regulation of phosphorylation
          response to peptide hormone
          protein kinase B signaling
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of glycogen biosynthetic process
          positive regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          positive regulation of insulin receptor signaling pathway
          negative regulation of insulin secretion
          insulin-like growth factor receptor signaling pathway
          protein heterooligomerization
          positive regulation of glucagon secretion
          negative regulation of somatostatin secretion
          positive regulation of glucose import in response to insulin stimulus
          signal transducer activity
          transmembrane receptor protein tyrosine kinase adaptor activity
          protein kinase C binding
          insulin receptor binding
          insulin-like growth factor receptor binding
          protein binding
          protein kinase binding
          protein domain specific binding
          SH2 domain binding
          phosphatidylinositol 3-kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P431109 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P149129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147898 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60122 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P149129 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P431109 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P149129 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P431109 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P149129 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P431109 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P431109 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81683 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39285 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56390 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929393 - -
          Affymetrix Rat Gene 1.0 ST Array 10929392 - -
          Affymetrix Rat Gene 1.0 ST Array 10929391 - -
          Affymetrix Rat Gene 1.1 ST Array 10929390 - -
          Rat Genome 230 2.0 Array 1369771_PM_at - -
          Rat Genome 230 2.0 Array 1369771_at - -
          Affymetrix HT Rat Focus Array 1369771_PM_at - -
          Affymetrix HT Rat Focus Array 1369771_at - -
          Affymetrix Rat Clariom S Assay PSR0900019168.rn.1 TC0900002338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019170.rn.1 TC0900002338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019171.rn.1 TC0900002338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019164.rn.1 TC0900002338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019166.rn.1 TC0900002338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019161.rn.1 TC0900002338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019169.rn.1 TC0900002338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019165.rn.1 TC0900002338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019160.rn.1 TC0900002338.rn.1 RTA TC
          Rat Expression Set 230 1369771_PM_at - -
          Rat Expression Set 230 1369771_at - -
          Affymetrix Rat Gene 1.0 ST Array 10929390 - -
          Rat Genome U34 Set X58375_at - -
          Rat Genome U34 Set rc_AA963662_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21886956 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360680 - -
          RatRef-12 Expression BeadChip ILMN_70074 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline