Search
Search
| Gene Symbol: Kcne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133603.2 | NP_598287.1 | 2 | 586 | 66 | |
| XM_006248023.2 | XP_006248085.1 | 2 | 1012 | 66 | ||
| XM_008768554.1 | XP_008766776.1 | 2 | 822 | 66 | ||
| GenBank mRNA | AF071003.1 | - | 2 | 427 | 66 | |
| BC089778.1 | - | 2 | 586 | 7301610 | 66 | |
Set Membership: |
|