Search
Search
| Gene Symbol: Rxfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008310.1 | NP_001008311.1 | 1 | 1327 | 156 | |
| GenBank mRNA | AY633763.1 | - | 1 | 1306 | 156 | |
| AY633764.1 | - | 1 | 1327 | 156 | ||
Set Membership: |
|