Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347198_g1
          See other CIRH1A GE Assays ›
          Gene Symbol:
          Cirh1a
          Gene Name:
          cirrhosis, autosomal recessive 1A (cirhin)
          Gene Aliases:
          Cirhin, Naic, Teg-292, Tex292
          Chromosome Location:
          Chr.19: 39087995 - 39116124 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02347198_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cirh1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009640.1 NP_001009640.1 16 - 17 1976 80
          XM_006255562.1 XP_006255624.1 16 - 17 2022 80
          XM_006255563.2 XP_006255625.1 16 - 17 2185 80
          XM_008772488.1 XP_008770710.1 15 - 16 1789 80
          GenBank mRNA BC088461.1 - 16 - 17 1976 7127580 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000530.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cirh1a

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          biological_process
          maturation of SSU-rRNA
          molecular_function
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P704824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129900 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P704824 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P704824 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P704824 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P704824 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83766 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807596 - -
          Affymetrix Rat Gene 1.0 ST Array 10807599 - -
          Affymetrix Rat Gene 1.0 ST Array 10807597 - -
          Affymetrix Rat Gene 1.0 ST Array 10807588 - -
          Affymetrix Rat Gene 1.0 ST Array 10807591 - -
          Affymetrix Rat Gene 1.0 ST Array 10807589 - -
          Affymetrix Rat Gene 1.0 ST Array 10807590 - -
          Affymetrix Rat Gene 1.0 ST Array 10807586 - -
          Affymetrix Rat Gene 1.0 ST Array 10807592 - -
          Affymetrix Rat Gene 1.0 ST Array 10807598 - -
          Affymetrix Rat Gene 1.0 ST Array 10807595 - -
          Affymetrix Rat Gene 1.0 ST Array 10807587 - -
          Affymetrix Rat Gene 1.0 ST Array 10807594 - -
          Affymetrix Rat Gene 1.0 ST Array 10782766 - -
          Affymetrix Rat Gene 1.0 ST Array 10807593 - -
          Affymetrix Rat Gene 1.0 ST Array 10807585 - -
          Affymetrix Rat Gene 1.0 ST Array 10807584 - -
          Affymetrix Rat Gene 1.0 ST Array 10807600 - -
          Affymetrix Rat Gene 1.1 ST Array 10807583 - -
          Affymetrix Rat Gene 1.1 ST Array 10782764 - -
          Rat Genome 230 2.0 Array 1389577_PM_at - -
          Rat Genome 230 2.0 Array 1389577_at - -
          Affymetrix HT Rat Focus Array 1389577_at - -
          Affymetrix HT Rat Focus Array 1389577_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900004667.rn.1 TC1900000530.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004665.rn.1 TC1900000530.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004666.rn.1 TC1900000530.rn.1 RTA TC
          Rat Expression Set 230 1389577_PM_at - -
          Rat Expression Set 230 1389577_at - -
          Affymetrix Rat Gene 1.0 ST Array 10807583 - -
          Rat Genome U34 Set rc_AI101160_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21455492 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349761 - -
          RatRef-12 Expression BeadChip ILMN_63252 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline