Search
Search
| Gene Symbol: Tor3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009683.1 | NP_001009683.1 | 3 - 4 | 637 | 117 | |
| GenBank mRNA | BC087678.1 | - | 3 - 4 | 637 | 7318816 | 117 |
| FQ226439.1 | - | 1 - 2 | 198 | 117 | ||
Set Membership: |
|