Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347895_m1
          See other CXADR GE Assays ›
          Gene Symbol:
          Cxadr
          Gene Name:
          coxsackie virus and adenovirus receptor
          Gene Aliases:
          Car1
          Chromosome Location:
          Chr.11: 16826298 - 16873564 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02347895_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxadr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053570.1 NP_446022.1 7 - 8 1087 141
          GenBank mRNA BC088313.1 - 7 - 8 1087 7324723 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000124.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxadr

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrion organization
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          heart development
          germ cell migration
          epithelial structure maintenance
          single organismal cell-cell adhesion
          neutrophil chemotaxis
          actin cytoskeleton reorganization
          homotypic cell-cell adhesion
          cell-cell junction organization
          gamma-delta T cell activation
          viral entry into host cell
          cardiac muscle fiber development
          defense response to virus
          negative regulation of cardiac muscle cell proliferation
          transepithelial transport
          AV node cell to bundle of His cell communication
          virus receptor activity
          receptor binding
          integrin binding
          beta-catenin binding
          PDZ domain binding
          identical protein binding
          cell adhesion molecule binding
          connexin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P013526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178708 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013526 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013526 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P013526 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P442813 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P716145 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85372 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94841 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23546 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8707 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10749986 - -
          Affymetrix Rat Gene 1.0 ST Array 10749992 - -
          Affymetrix Rat Gene 1.0 ST Array 10749985 - -
          Affymetrix Rat Gene 1.0 ST Array 10749987 - -
          Affymetrix Rat Gene 1.0 ST Array 10749989 - -
          Affymetrix Rat Gene 1.0 ST Array 10749988 - -
          Affymetrix Rat Gene 1.0 ST Array 10749990 - -
          Affymetrix Rat Gene 1.0 ST Array 10749984 - -
          Affymetrix Rat Gene 1.0 ST Array 10749993 - -
          Affymetrix Rat Gene 1.1 ST Array 10749983 - -
          Rat Genome 230 2.0 Array 1384816_PM_at - -
          Rat Genome 230 2.0 Array 1384816_at - -
          Affymetrix HT Rat Focus Array 1384816_PM_at - -
          Affymetrix HT Rat Focus Array 1384816_at - -
          Affymetrix Rat Clariom S Assay PSR1100000421.rn.1 TC1100000124.rn.1 RTA TC
          Rat Expression Set 230 1384816_at - -
          Rat Expression Set 230 1384816_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10749983 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22130228 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352802 - -
          RatRef-12 Expression BeadChip ILMN_61643 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline