Search
Search

Osgepl1| Gene Symbol: Osgepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024787.1 | NP_001019958.1 | 7 - 8 | 1389 | 93 | |
| XM_006244893.2 | XP_006244955.1 | 7 - 8 | 1438 | 93 | ||
| XM_006244894.2 | XP_006244956.1 | 7 - 8 | 1402 | 93 | ||
| GenBank mRNA | BC097950.1 | - | 7 - 8 | 1389 | 7389045 | 93 |
Set Membership: |
|