Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02397280_mH
          See other UGT1A9 GE Assays ›
          Gene Symbol:
          Ugt1a9
          Gene Name:
          UDP glucuronosyltransferase 1 family, polypeptide A9
          Gene Aliases:
          Ugt1a10, Ugt1a11
          Chromosome Location:
          Chr.9: 95161157 - 95302822 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02397280_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt1a9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201425.2 NP_958828.1 1 - 2 856 130
          GenBank mRNA AY435129.1 - 1 - 2 856 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001092.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugt1a9

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          biological_process
          drug metabolic process
          negative regulation of fatty acid metabolic process
          negative regulation of transferase activity
          flavone metabolic process
          cellular glucuronidation
          flavonoid glucuronidation
          xenobiotic glucuronidation
          negative regulation of cellular glucuronidation
          retinoic acid binding
          molecular_function
          enzyme inhibitor activity
          glucuronosyltransferase activity
          enzyme binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P363385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65608 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34182 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101539 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12740 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P363385 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P363385 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P363385 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17845 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45134 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8182 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51590 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50909 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32020 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21359 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93650 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5296 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925182 - -
          Affymetrix Rat Gene 1.0 ST Array 10925179 - -
          Affymetrix Rat Gene 1.0 ST Array 10925177 - -
          Affymetrix Rat Gene 1.0 ST Array 10925184 - -
          Affymetrix Rat Gene 1.0 ST Array 10925178 - -
          Affymetrix Rat Gene 1.0 ST Array 10925159 - -
          Affymetrix Rat Gene 1.0 ST Array 10925180 - -
          Affymetrix Rat Gene 1.0 ST Array 10925181 - -
          Affymetrix Rat Gene 1.0 ST Array 10925176 - -
          Affymetrix Rat Gene 1.0 ST Array 10925183 - -
          Affymetrix Rat Gene 1.1 ST Array 10925158 - -
          Rat Genome 230 2.0 Array 1387759_s_at - -
          Rat Genome 230 2.0 Array 1370613_PM_s_at - -
          Rat Genome 230 2.0 Array 1370613_s_at - -
          Rat Genome 230 2.0 Array 1387759_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0900009373.rn.1 TC0900001092.rn.1 RTA TC
          Rat Expression Set 230 1370613_s_at - -
          Rat Expression Set 230 1387759_PM_s_at - -
          Rat Expression Set 230 1370613_PM_s_at - -
          Rat Expression Set 230 1387759_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925158 - -
          Rat Genome U34 Set S56937_s_at - -
          Rat Genome U34 Set J02612mRNA_s_at - -
          Rat Genome U34 Set J05132_s_at - -
          Rat Genome U34 Set D83796_s_at - -
          Rat Genome U34 Set J05132_s_at - -
          Rat Genome U34 Set J02612mRNA_s_at - -
          Rat Genome U34 Set D83796_s_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline