Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02397545_g1
          See other EAR11 GE Assays ›
          Gene Symbol:
          Ear11 Ear1 LOC100361866
          Gene Name:
          eosinophil-associated, ribonuclease A family, member 11
          eosinophil-associated, ribonuclease A family, member 1
          eosinophil cationic protein-like
          Gene Aliases:
          EAR, ECP, R1, R7, Rnase3
          Ear3, Raf4
          -
          Chromosome Location:
          Chr.15: 28216744 - 28217614 on Build Rnor_6.0
          Chromosome Location:
          Chr.15: 28260129 - 28261049 on Build Rnor_6.0
          Chromosome Location:
          Chr.15: 28240548 - 28241470 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn02397545_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ear11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138902.1 NP_620257.1 650 124
          GenBank mRNA D88586.1 - 650 124
          Gene Symbol: Ear1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002728193.3 XP_002728239.1 2 696 124
          Gene Symbol: LOC100361866
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002728192.3 XP_002728238.1 2 699 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ear11
          Click here to find siRNAs for Gene Ear1
          Click here to find siRNAs for Gene LOC100361866

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          antibacterial humoral response
          defense response to Gram-positive bacterium
          nucleic acid phosphodiester bond hydrolysis
          nucleic acid binding
          endonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P358160 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P358160 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P358160 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P358160 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P358160 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783211 - -
          Affymetrix Rat Gene 1.0 ST Array 10783212 - -
          Affymetrix Rat Gene 1.0 ST Array 10786116 - -
          Affymetrix Rat Gene 1.0 ST Array 10786117 - -
          Affymetrix Rat Gene 1.1 ST Array 10783210 - -
          Affymetrix Rat Gene 1.1 ST Array 10786115 - -
          Rat Genome 230 2.0 Array 1370777_PM_at - -
          Rat Genome 230 2.0 Array 1370777_at - -
          Affymetrix HT Rat Focus Array 1370777_at - -
          Affymetrix HT Rat Focus Array 1370777_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500009947.rn.1 TC1500001573.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009948.rn.1 TC1500001573.rn.1 RTA TC
          Rat Expression Set 230 1370777_PM_at - -
          Rat Expression Set 230 1370777_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783210 - -
          Affymetrix Rat Gene 1.0 ST Array 10786115 - -
          Rat Genome U34 Set D88586_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22031694 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363762 - -
          RatRef-12 Expression BeadChip ILMN_69699 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0