Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02586393_s1
          See other HIST3H2A GE Assays ›
          Gene Symbol:
          Hist3h2a
          Gene Name:
          histone cluster 3, H2a
          Gene Aliases:
          H2a
          Chromosome Location:
          Chr.10: 45297603 - 45299155 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02586393_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist3h2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021840.2 NP_068612.2 1 1367 158
          GenBank mRNA BC099140.1 - 1 1367 7457173 158

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hist3h2a

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome disassembly
          UV-damage excision repair
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P531971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22196 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P531971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P531971 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P531971 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P146616 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P396522 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63717 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733941 - -
          Affymetrix Rat Gene 1.1 ST Array 10733940 - -
          Rat Genome 230 2.0 Array 1371765_at - -
          Rat Genome 230 2.0 Array 1381208_at - -
          Rat Genome 230 2.0 Array 1371765_PM_at - -
          Rat Genome 230 2.0 Array 1382473_PM_at - -
          Rat Genome 230 2.0 Array 1381208_PM_at - -
          Rat Genome 230 2.0 Array 1382473_at - -
          Affymetrix HT Rat Focus Array 1369892_PM_at - -
          Affymetrix HT Rat Focus Array 1381208_PM_at - -
          Affymetrix HT Rat Focus Array 1371765_at - -
          Affymetrix HT Rat Focus Array 1369892_at - -
          Affymetrix HT Rat Focus Array 1381208_at - -
          Affymetrix HT Rat Focus Array 1371765_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000005920.rn.1 TC1000000710.rn.1 RTA TC
          Rat Expression Set 230 1371765_at - -
          Rat Expression Set 230 1371765_PM_at - -
          Rat Expression Set 230 1381208_PM_at - -
          Rat Expression Set 230 1381208_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733940 - -
          Rat Genome U34 Set rc_AA944410_at - -
          Rat Genome U34 Set rc_AA850895_at - -
          Rat Genome U34 Set rc_AA998848_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21229926 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline