Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02586536_g1
          See other CYTH2 GE Assays ›
          Gene Symbol:
          Cyth2
          Gene Name:
          cytohesin 2
          Gene Aliases:
          Arno, Pscd2, Sec7, Sec7B
          Chromosome Location:
          Chr.1: 101796349 - 101803204 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02586536_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyth2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053911.2 NP_446363.1 11 - 12 1146 81
          XM_006228964.2 XP_006229026.1 10 - 11 1161 81
          XM_008759304.1 XP_008757526.1 10 - 11 1637 81
          GenBank mRNA FQ229744.1 - 9 - 10 864 81
          U83896.1 - 11 - 12 1150 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005408.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyth2

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle-mediated transport
          regulation of cell adhesion
          regulation of ARF protein signal transduction
          positive regulation of GTPase activity
          negative regulation of dendrite development
          ARF guanyl-nucleotide exchange factor activity
          protein binding
          lipid binding
          inositol 1,4,5 trisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111920 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12977 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12977 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721911 - -
          Affymetrix Rat Gene 1.0 ST Array 10721917 - -
          Affymetrix Rat Gene 1.0 ST Array 10721908 - -
          Affymetrix Rat Gene 1.0 ST Array 10721916 - -
          Affymetrix Rat Gene 1.0 ST Array 10721906 - -
          Affymetrix Rat Gene 1.0 ST Array 10721915 - -
          Affymetrix Rat Gene 1.0 ST Array 10721912 - -
          Affymetrix Rat Gene 1.0 ST Array 10721905 - -
          Affymetrix Rat Gene 1.0 ST Array 10721910 - -
          Affymetrix Rat Gene 1.0 ST Array 10721913 - -
          Affymetrix Rat Gene 1.0 ST Array 10721907 - -
          Affymetrix Rat Gene 1.0 ST Array 10721914 - -
          Affymetrix Rat Gene 1.0 ST Array 10721909 - -
          Affymetrix Rat Gene 1.1 ST Array 10721904 - -
          Rat Genome 230 2.0 Array 1367727_at - -
          Rat Genome 230 2.0 Array 1367727_PM_at - -
          Affymetrix HT Rat Focus Array 1367727_at - -
          Affymetrix HT Rat Focus Array 1367727_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100044242.rn.1 TC0100005408.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100044240.rn.1 TC0100005408.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100044243.rn.1 TC0100005408.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100044241.rn.1 TC0100005408.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100044239.rn.1 TC0100005408.rn.1 RTA TC
          Rat Expression Set 230 1367727_PM_at - -
          Rat Expression Set 230 1367727_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721904 - -
          Rat Genome U34 Set U83896_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21528374 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373520 - -
          RatRef-12 Expression BeadChip ILMN_62102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline