Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02606541_m1
          See other ESRRB GE Assays ›
          Gene Symbol:
          Esrrb
          Gene Name:
          estrogen-related receptor beta
          Gene Aliases:
          Err2, Errb
          Chromosome Location:
          Chr.6: 110300942 - 110456061 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02606541_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Esrrb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008516.2 NP_001008516.2 4 - 5 787 53
          XM_006240374.2 XP_006240436.1 5 - 6 1249 53
          XM_006240375.2 XP_006240437.1 5 - 6 1250 53
          XM_008764759.1 XP_008762981.1 5 - 6 2485 53
          GenBank mRNA AY383731.1 - 4 - 5 787 53

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001210.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Esrrb

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          trophectodermal cellular morphogenesis
          trophectodermal cell proliferation
          embryonic placenta development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          stem cell population maintenance
          steroid hormone mediated signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          transcription coactivator activity
          steroid binding
          transcription factor binding
          zinc ion binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P115811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45624 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115811 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115811 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P115811 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P381969 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70004 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7764 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39907 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886123 - -
          Affymetrix Rat Gene 1.0 ST Array 10886125 - -
          Affymetrix Rat Gene 1.0 ST Array 10886127 - -
          Affymetrix Rat Gene 1.0 ST Array 10886126 - -
          Affymetrix Rat Gene 1.0 ST Array 10886124 - -
          Affymetrix Rat Gene 1.0 ST Array 10886122 - -
          Affymetrix Rat Gene 1.1 ST Array 10886121 - -
          Affymetrix Rat Clariom S Assay PSR0600008617.rn.1 TC0600001210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008618.rn.1 TC0600001210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008616.rn.1 TC0600001210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008619.rn.1 TC0600001210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008612.rn.1 TC0600001210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008615.rn.1 TC0600001210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008611.rn.1 TC0600001210.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10886121 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372917 - -
          RatRef-12 Expression BeadChip ILMN_55858 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline