Search
Search

LOC500990| Gene Symbol: LOC500990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037364.1 | NP_001032441.1 | 5 - 6 | 580 | 70 | |
| XM_008766163.1 | XP_008764385.1 | 6 - 7 | 796 | 70 | ||
| XM_008766164.1 | XP_008764386.1 | 6 - 7 | 796 | 70 | ||
| XM_008766165.1 | XP_008764387.1 | 6 - 7 | 796 | 70 | ||
| XM_008766166.1 | XP_008764388.1 | 6 - 7 | 796 | 70 | ||
| XM_008766167.1 | XP_008764389.1 | 6 - 7 | 793 | 70 | ||
| XM_008766168.1 | XP_008764390.1 | 6 - 7 | 807 | 70 | ||
| XM_008766169.1 | XP_008764391.1 | 5 - 6 | 713 | 70 | ||
| XM_008766170.1 | XP_008764392.1 | 5 - 6 | 668 | 70 | ||
| GenBank mRNA | BC105843.1 | - | 5 - 6 | 580 | 7455643 | 70 |
Set Membership: |
|