Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02769676_s1
          See other MYL2 GE Assays ›
          Gene Symbol:
          Myl2
          Gene Name:
          myosin, light chain 2, regulatory, cardiac, slow
          Gene Aliases:
          Mlc-2, Mlc2
          Chromosome Location:
          Chr.12: 39951863 - 39959065 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02769676_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001035252.2 NP_001030329.2 7 528 97
          GenBank mRNA BC126064.1 - 7 528 7190683 97
          M11851.1 - 7 537 97
          X07314.1 - 7 521 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myl2

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          heart morphogenesis
          heart development
          post-embryonic development
          negative regulation of cell growth
          muscle cell fate specification
          muscle fiber development
          cardiac myofibril assembly
          ventricular cardiac muscle tissue morphogenesis
          heart contraction
          cardiac muscle contraction
          actin monomer binding
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P397163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P397163 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P397163 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P397163 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P397163 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700292577 - -
          Agilent Rat cDNA Microarray (G4105A) 700292577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762081 - -
          Affymetrix Rat Gene 1.0 ST Array 10762080 - -
          Affymetrix Rat Gene 1.0 ST Array 10762068 - -
          Affymetrix Rat Gene 1.1 ST Array 10762067 - -
          Rat Genome 230 2.0 Array 1388200_PM_at - -
          Rat Genome 230 2.0 Array 1388200_at - -
          Rat Genome 230 2.0 Array 1388293_at - -
          Rat Genome 230 2.0 Array 1388293_PM_at - -
          Affymetrix HT Rat Focus Array 1388293_at - -
          Affymetrix HT Rat Focus Array 1388200_at - -
          Affymetrix HT Rat Focus Array 1388293_PM_at - -
          Affymetrix HT Rat Focus Array 1388200_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200013670.rn.1 TC1200001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013669.rn.1 TC1200001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013665.rn.1 TC1200001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013664.rn.1 TC1200001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013673.rn.1 TC1200001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013667.rn.1 TC1200001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013671.rn.1 TC1200001604.rn.1 RTA TC
          Rat Expression Set 230 1388293_PM_at - -
          Rat Expression Set 230 1388200_PM_at - -
          Rat Expression Set 230 1388293_at - -
          Rat Expression Set 230 1388200_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762067 - -
          Rat Genome U34 Set X07314cds_at - -
          Rat Genome U34 Set M11851_at - -
          Rat Genome U34 Set X07314cds_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21365535 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline