Search
Search
| Gene Symbol: Gbgt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB081649.1 | - | 7 | 865 | 144 | |
| AB081650.1 | - | 7 | 865 | 144 | ||
| AF296762.1 | - | 896 | 144 | |||
| AF469946.1 | - | 6 | 790 | 144 | ||
Set Membership: |
|