Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03062000_g1
          See other VOM2R69 GE Assays ›
          Gene Symbol:
          Vom2r69
          Gene Name:
          vomeronasal 2 receptor, 69
          Gene Aliases:
          RGD1564750
          Chromosome Location:
          Chr.14: 1007495 - 1020982 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03062000_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vom2r69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099468.1 NP_001092938.1 2 - 3 509 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174598 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112954 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112954 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112954 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P194374 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774927 - -
          Affymetrix Rat Gene 1.0 ST Array 10774929 - -
          Affymetrix Rat Gene 1.0 ST Array 10774928 - -
          Affymetrix Rat Gene 1.0 ST Array 10774926 - -
          Affymetrix Rat Gene 1.0 ST Array 10774925 - -
          Affymetrix Rat Gene 1.1 ST Array 10774924 - -
          Affymetrix Rat Clariom S Assay PSR1400000108.rn.1 TC1400000015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000107.rn.1 TC1400000015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000109.rn.1 TC1400000015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000104.rn.1 TC1400000015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000105.rn.1 TC1400000015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000106.rn.1 TC1400000015.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10774924 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362789 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline