Search
Search
| Gene Symbol: Pbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100681.1 | NP_001094151.1 | 3 - 4 | 763 | 84 | |
| NM_001134862.1 | NP_001128334.1 | 3 - 4 | 933 | 84 | ||
| GenBank mRNA | BC161983.1 | - | 3 - 4 | 763 | 9030389 | 84 |
Set Membership: |
|